Skip to Content
Merck
All Photos(1)

Key Documents

690014X

Avanti

Solvent Mixture 20:9:1

Avanti Research - A Croda Brand

Synonym(s):

PIP Mixture

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$257.00

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$257.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

chloroform:methanol:water

packaging

pkg of 4 × 1 mL each (690014X-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

storage temp.

2-30°C

Packaging

5 mL Clear Glass Sealed Ampules, packaged as 4 × 1 mL (690014X-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service