Skip to Content
Merck
All Photos(1)

Key Documents

610032

Avanti

Plunger Assembly (1000 μL)

Avanti Research - A Croda Brand

Synonym(s):

Plunger Assembly, Plunger Assembly for 1000μL Hamilton Syringe

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$67.50

$67.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$67.50

About This Item

UNSPSC Code:
12352211
NACRES:
NB.03

$67.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Replacement Part for Avanti′s Mini-Extruder.

Watch the video to learn more about the Avanti Research Mini-Extruder.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service