Skip to Content
Merck
All Photos(1)

Key Documents

610005

Avanti

PC Membranes 0.1um

Avanti Research - A Croda Brand

Synonym(s):

Extruder PC Membrane, Polycarbonate Membranes 0.1μm 19mm (100/pk)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$664.00

$664.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$664.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

$664.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

PC Membranes are filter membranes possessing smooth and flat surfaces. They consist of track-etched uniform cylindrical pores.[1]
Polycarbonate Membrane for Avanti′s Mini-Extruder.

Watch the video to learn more about the Avanti Research Mini-Extruder

Application

PC Membranes 0.1um has been used to extrude unilamellar liposomes[2][3] and multilamellar vesicles.[4]

Biochem/physiol Actions

PC Membrane provides fine mechanical properties, chemical stability, and non-auto fluorescent properties.[5] It is extensively used for the identification of various bacteria in immunofluorescent direct count method.[5]

Components

Whatman® Nuclepore polycarbonate hydrophilic membrane 800309

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoosoo Yang et al.
Frontiers in immunology, 9, 725-725 (2018-04-27)
Vesicle-associated V-soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) proteins and target membrane-associated T-SNAREs (syntaxin 4 and SNAP-23) assemble into a core trans-SNARE complex that mediates membrane fusion during mast cell degranulation. This complex plays pivotal roles at various stages of
Response of MG63 osteoblast-like cells onto polycarbonate membrane surfaces with different micropore sizes
Lee SJ, et al.
Biomaterials, 25(19), 4699-4707 (2004)
Farzin Haque et al.
Nature protocols, 8(2), 373-392 (2013-01-26)
Over the past decade, nanopores have rapidly emerged as stochastic biosensors. This protocol describes the cloning, expression and purification of the channel of the bacteriophage phi29 DNA-packaging nanomotor and its subsequent incorporation into lipid membranes for single-pore sensing of double-stranded
Use of the supported membrane tube assay system for real time analysis of membrane fission reactions
Dar S, et al.
Nature Protocols, 12(2), 390-390 (2017)
Highly sensitive detection of Salmonella typhi using surface aminated polycarbonate membrane enhanced-ELISA
Jain S, et al.
Biosensors And Bioelectronics, 31(1), 37-43 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service