Skip to Content
Merck
All Photos(2)

Key Documents

330277W

Avanti

Glycocholic acid-d4

Avanti Research - A Croda Brand

Synonym(s):

5ß-cholanic acid-3α,7α,12α-triol N-(carboxymethyl)-amide-d4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$274.00

$274.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$274.00

About This Item

Empirical Formula (Hill Notation):
C26H39D4NO6
CAS Number:
Molecular Weight:
469.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

$274.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Assay

99% (MS)

form

methanol solution

packaging

package of 1 × 100 μg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

100 μg/mL in methanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

N(CC(=O)O)C(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@@H](C([C@@H](C(C4)([2H])[2H])O)([2H])[2H])C[C@H]3O)C)C[C@@H]2O)CC1)C)C

InChI

1S/C26H43NO6/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33)/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-/m1/s1/i8D2,10D2

Packaging

2 mL Amber Glass Sealed Ampule (330277W-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service