Skip to Content
Merck
All Photos(1)

Key Documents

809675

Sigma-Aldrich

Deoxycholic-2,2,4,4,11,11-d6 acid

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,420.00

$1,420.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$1,420.00

About This Item

Empirical Formula (Hill Notation):
C24D6H34O4
Molecular Weight:
398.61
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,420.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

[2H]C1([2H])[C@@H](O)C([2H])([2H])C(CCC2C3C([2H])([2H])[C@H](O)[C@@]4(C)C2CC[C@H]4[C@@H](CCC(O)=O)C)[C@]3(C)C1

InChI

1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15?,16-,17?,18?,19?,20?,21+,23+,24-/m1/s1/i10D2,12D2,13D2

InChI key

KXGVEGMKQFWNSR-UEKQVLGMSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huasheng Yu et al.
eLife, 8 (2019-09-11)
Patients with liver diseases often suffer from chronic itch, yet the pruritogen(s) and receptor(s) remain largely elusive. Here, we identify bile acids as natural ligands for MRGPRX4. MRGPRX4 is expressed in human dorsal root ganglion (hDRG) neurons and co-expresses with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service