Z767301
Scienceware® portable glove box system with gas ports
large size neoprene gloves with clamping rings
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H50028-2001
General description
Lightweight, self-contained portable isolation cabinet has two gas ports on the back of the box that allow users to create gas specific work environments. A large acrylic window with 6 in. circular glove ports permits safe viewing of operations, and the 8 in. diameter side port allows easy insertion of material and equipment.
- Economical containment unit ideal for quick set-up anywhere
- Gas ports with tapered fitting accept 1/4 in. to 3/8 in. I.D. tubing and have easy turning PTFE stopcocks
- Large size, 22 in. long (20 in. installed) neoprene gloves and 6 in. circular port clamping rings (included)
- Round corners throughout for easy cleaning
- Body molded from LDPE
- Overall dimensions: 68.5 W x 33D x 55 cm H (27 x 13 x 22 in.); gas ports protrude 2.6 in. from the back of the box
- Internal volume: 86.6 L
- Shipping weight: 17.2 kg (38 lbs)
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service