Z765252
Tygon® formula E-3603 laboratory tubing
I.D. × O.D. × wall thickness 1 in. × 2 in. × 1/2 in.
Synonym(s):
DEHP tubing alternative, chemical resistant tubing, laboratory tubing, vacuum tubing
Select a Size
$5,280.00
Select a Size
About This Item
$5,280.00
Recommended Products
packaging
pkg of 50 feet
manufacturer/tradename
Tygon
I.D. × O.D. × wall thickness
1 in. × 2 in. × 1/2 in.
Looking for similar products? Visit Product Comparison Guide
General description
TYGON® E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.
TYGON® E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.
Features/Benefits:
- Alternative to DEHP tubing
- New formulation delivers the same excellent performance
- Contains no BPA
- Outstanding chemical resistance
- Lot-to-lot consistency for reproducible results
- Increases productivity in peristaltic pumps - outlasts other clear tubing 2 to 1
- Smooth, polished inner bore
- Non-oxidizing and non-contaminating
- Slips easily over fittings and grips securely
- Coils marked at 1-foot intervals for easy measuring
- Meets FDA criteria
Typical Applications:
- General laboratory
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain lines
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service