Skip to Content
Merck
All Photos(1)

Key Documents

Z757020

Kern sample plates for DBS moisture analyzer

aluminum, pack of 80 ea

Synonym(s):

moisture sensor, moisture tester

Sign Into View Organizational & Contract Pricing

Select a Size

80 EA
$74.40

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
80 EA
$74.40

About This Item

UNSPSC Code:
41111502
NACRES:
NB.71
material:
aluminum

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

aluminum

sterility

non-sterile

packaging

pack of 80 ea

manufacturer/tradename

Kern MLBA01A

color

none

General description

Aluminimum sample plates, 90 mm diameter, pack of 80, for use with the Kern DAB moisture analyzer.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service