Skip to Content
Merck
All Photos(1)

Documents

Z221872

Disposable weighing pan

aluminum, diam. × H 100 mm × 8 mm, pack of 50 ea

Synonym(s):

Weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71

material

aluminum
circles

packaging

pack of 50 ea

manufacturer/tradename

Sigma-Aldrich

diam. × H

100 mm × 8 mm

dish diam.

100 mm

General description

Oil-free.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service