Skip to Content
Merck
All Photos(1)

Key Documents

Z743859

ART® Non-Filtered Tip

HLT 5000, size 5 mL, micropoint bulk pack, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 250 EA
$793.00

$793.00


Estimated to ship onApril 17, 2025



Select a Size

Change View
10 X 250 EA
$793.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
size:
5 mL
sterility:
non-sterile
feature:
HLT 5000
micropoint bulk pack
packaging:
pkg of 10X250 ea
manufacturer/tradename:
Molecular BioProducts 3896

$793.00


Estimated to ship onApril 17, 2025


sterility

non-sterile

feature

HLT 5000
micropoint bulk pack

packaging

pkg of 10X250 ea

manufacturer/tradename

Molecular BioProducts 3896

technique(s)

PCR: suitable

size

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

ART™ Non-filtered Hinged Rack Pipette Tips, which are made for the way you work, allows you to access tips with just one hand. You need not be concerned about your tips spilling on the bench or floor anymore. In just three simple steps, you can go from hinged to lift-off and maximize your productivity. Just unhinge, turn, and realign the cover to have a conventional lift-off when necessary. These tips are appropriate if contamination is not a concern.

Features and Benefits

  • Complete selection of non-filtered and specialty tips provides ART quality, performance, and reliability.
  • Racks allow saving time by offering quick and easy access to pipette tips.
  • The front latch closes and locks to provide safe and secure storage

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service