Skip to Content
Merck
All Photos(1)

Key Documents

Z743696

ART® Barrier Tip

ART 1000 SoftFit, size 1000 μL, individually wrapped, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$261.00

$261.00


Estimated to ship onApril 11, 2025



Select a Size

Change View
500 EA
$261.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
size:
1000 μL
sterility:
sterile
feature:
individually wrapped
ART 1000 SoftFit
packaging:
pkg of 500 ea
manufacturer/tradename:
Molecular BioProducts 2279IW

$261.00


Estimated to ship onApril 11, 2025


sterility

sterile

feature

individually wrapped
ART 1000 SoftFit

packaging

pkg of 500 ea

manufacturer/tradename

Molecular BioProducts 2279IW

technique(s)

PCR: suitable

size

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

ART™ Barrier Reload System helps to perform applications precisely and consistently. This system ensures both accuracy and protection while maintaining sterility. With the ability to stack tips due to offset wells, it minimizes waste and optimizes benchtop and storage space.

Features and Benefits

  • ART barrier tips ensure complete protection against aerosols and liquid contamination.
  • The tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
  • Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
  • Blister sleeve guide rails facilitate smooth transfer of tip inserts into the rack.
  • An outer sleeve shields tips from external contamination during the refilling of inserts.
  • The tip packaging option helps reduce plastic waste.
  • The rigid insert design ensures reliable tip loading.
  • Lot number and description are printed on every insert for easy tracking and identification.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service