Key Documents
Z743189
Julabo® bath covers
with built in heat exchanger to fit CORIO CD-BC4/BC6
Select a Size
1 EA
$959.00
$959.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$959.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$959.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970240
General description
Bath cover with built-in heat exchanger. To use the heat exchanger, a hose with an inner diameter of 10/12 mm is required.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)