Key Documents
Z743184
Julabo® bath covers
lift-up transparent cover to fit Pura 30
Select a Size
1 EA
$732.00
$732.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$732.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$732.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970584
General description
The hinged cover for use with the PURA 30 water bath reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)