Z743014
Julabo® CORIO™ CD heating circulator with open bath
CD-B17, capacity 13 L (17 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, Schuko plug
All Photos(1)
About This Item
Recommended Products
material
stainless steel bath
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9012417
temperature measuring range
20-150 °C
capacity
13 L (17 L)
external W × L × H
38 cm × 40 cm × 47 cm
opening W × L × H
30 cm × 18 cm × 20 cm
General description
The Open Heating Bath Circulators of the new CORIO series feature high-quality bath tanks made of stainless steel and pump connections.
Your advantages
Cooling coil and bath cover available as optional accessories
Your advantages
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- USB connection
- High-quality bath tanks made of stainless steel
- Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.