Key Documents
Z740159
LifeSep magnetic separator unit
96F, for 96 well tray
Synonym(s):
cell sorting magnetic seperation unit
Select a Size
1 EA
$1,850.00
$1,850.00
Estimated to ship onMarch 20, 2025
Select a Size
Change View
1 EA
$1,850.00
About This Item
UNSPSC Code:
41100000
NACRES:
NA.56
$1,850.00
Estimated to ship onMarch 20, 2025
feature
for 96 well tray
manufacturer/tradename
Dexter Magnetic Technologies 2501008-1
General description
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
Choose from one of the most recent versions:
Certificates of Analysis (COA)