Z742224
Ohaus® Scout® SPX Balance
Model SPX2202, weighing capacity 2200 g, resolution: 0.01 g
Synonym(s):
laboratory balance, weighing balance
About This Item
Recommended Products
AC/DC input
120 V AC, US 3-pin plug
material
stainless steel platform
Agency
suitable for EPA 200.7
feature
CSA approved
display LCD
manufacturer/tradename
Ohaus® 30253023
weighing capacity measuring range
2200 g, resolution: 0.01 g
platform W × D
170 mm × 140 mm
battery power
4 AA, 80 hr life
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Bright Backlit LCD & 4 Button Control Enable Efficient Operation and Ease of Use
Combined with a simplified 4 button operation, the Scout′s large backlit display increases readability in low light working conditions, and is easier to view from a distance.
- Bolstered by Superior Overload Protection and Impact-Resistant Pan Support
Integrated superior overload protection and impact-resistant pan support safeguard against shock and overloading the balance in rugged and demanding environments.
- Better Performance and Higher Capacities Widen the Range of Weighing Applications
Stabilization time as fast as 1 sec and advanced weighing technology, lead to improved productivity in your facility. Increased capacity unlocks a wider range of weighing applications.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service