Key Documents
Z740620
Sigma-Aldrich
Aldrich® Essentials petri dish
soda lime glass, autoclavable
Select a Size
24 EA
$202.00
$202.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
24 EA
$202.00
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
$202.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
material
soda-lime glass
sterility
non-sterile
feature
autoclavable
lid
packaging
pack of 24 ea
dish capacity
~100 mL
binding type
non-treated surface
General description
Clear, soft flint glass Petri dishes.Bottom and top matched. Reusable soda lime glass, can be autoclaved.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)