Z694169
Troemner Lab-Frame standard kit with aluminum rods
X-large, 48 in. x 72 in.
Synonym(s):
laboratory clamps, laboratory support rods
Select a Size
$1,710.00
Select a Size
About This Item
$1,710.00
Recommended Products
size
XL
manufacturer/tradename
Troemner 916176
frame L × W
48 in. × 72 in.
rod diam.
13 mm
Looking for similar products? Visit Product Comparison Guide
General description
Components:
- 2 in. (51 mm) rods (qty 6)
- 12 in. (305 mm) rods (qty 6)
- 24 in. (610 mm) rods (qty 2)
- 48 in. (1219 mm) rods (qty 7)
- 72 in. (1829 mm) rods (qty 5)
- S-connectors (qty 45)
- Rod end connectors (qty 4)
- Lab-frame feet (qty 8)
- Traditional center-less ground aluminum
- Non-corroding fiberglass
- Tough stainless steel
A range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service