Key Documents
Z693782
LabJaws lattice rod S-connector
stainless steel
Synonym(s):
clamp holders, frame connectors
Select a Size
1 EA
$86.60
$86.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$86.60
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
$86.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
stainless steel
manufacturer/tradename
Troemner 916355
jaw capacity
0-13 mm
General description
Clamps two lattice rods at a 90° angle and fetures separate adjustment screws for each rod location.
Legal Information
LabJaws is a trademark of Henry Troemner LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)