Documents
Z693812
LabJaws multi-rod connector
nickel plated
Synonym(s):
clamp holders, frame connectors
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
nickel plated
manufacturer/tradename
Troemner 916131
jaw capacity
0-13 mm
Related Categories
General description
Maximum adjustment capability within one connector. Two adjustment rods fit into the oval hole in the connector, preventing turning when tightened. Side and front 13 mm holes allow creation of a variety of configurations.
Legal Information
LabJaws is a trademark of Henry Troemner LLC