Key Documents
Z684562
Ace Rotary Evaporator Vial Adapter Kit
Chemraz O-ring, adapter joint: ST/NS 24/40
Select a Size
1 EA
$1,850.00
$1,850.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,850.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$1,850.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
Chemraz O-ring
Joint
joint: ST/NS adapter24/40
description
PTFE adapters with #15 Ace-Thred to GPI vial thread, sizes 8-425, 13-425, 15-425, 18-400, 20-400, 22-400, 24-410
manufacturer/tradename
Ace Glass 13290-59
General description
PTFE connecting adapters and glass conversion adapters for connecting GPI threaded vials to a rotary evaporator vapor tube or trap. Both are suitable for vacuum work. PTFE connecting adapters are available with FETFE® or Chemraz® O-rings. Order Chemraz if you are using methylene chloride or acetone.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Chemraz is a registered trademark of Greene, Tweed of Delaware, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)