Key Documents
Z683205
ALNOCHROMIX safety vent caps
fits 1 gal or 2.5 L glass bottles
Select a Size
3 EA
$219.00
$219.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
3 EA
$219.00
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
$219.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Alconox, Inc. 2533
General description
When storing ALNOCHROMIX solution in the Sulfuric Acid glass bottle (1 gallon or 2.5 liter), our Safety Vent Cap will prevent pressure build-up by permitting gases to escape automatically as internal pressure increases.
Legal Information
ALNOCHROMIX is a trademark of Alconox, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)