Merck
All Photos(5)
Key Documents
Z680494
Duran® GL 45 connection system caps and accessories
insert for hose connection screw cap GL 14, 6.0 mm
Synonym(s):
1129818, Duran Cap Accessories, GL 45 Connection Caps
Select a Size
1 EA
$86.10
$86.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$86.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$86.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
white PTFE/silicone
manufacturer/tradename
DURAN® 1129818
parameter
-50-230 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Features and Benefits
  • Manufactured from PP, PTFE, silicone
  • Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
  • Safe and secure liquid transfer into or out of laboratory bottles
  • Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
  • Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
  • Closed-top red PBT GL 14 cap for sealing unused ports
  • Sterile pressure equilisation is possible with pressure compensation set
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)