Skip to Content
Merck
All Photos(1)

Documents

Z678473

Krytox® XP vacuum oil

grade, 1506

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161501
NACRES:
NB.79

mol wt

2160

feature

grade 1506

manufacturer/tradename

Du Pont

viscosity number

(20 °C) (62 mm2/s)

density

1.88 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

Krytox XP oils contain a soluble additive to prevent rust. This additive enhances the
performance of Krytox vacuum pump fluids, giving them improved performance
properties. The long-term antirust properties repel moisture, providing extra
protection from corrosion of metal parts and bearing surfaces.

  • Rust preventing
  • Nonflammable
  • Nonreactive
  • Free from acetal groups
  • Recyclable

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service