Skip to Content
Merck
All Photos(2)

Key Documents

Z676632

ART® low retention barrier tips

Tip design ART 20P, Reload insert

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 960 EA
$1,110.00

$1,110.00


Estimated to ship onMay 22, 2025



Select a Size

Change View
5 X 960 EA
$1,110.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
20 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
Reload insert
Tip design ART 20P
packaging:
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2149P-05-RI

$1,110.00


Estimated to ship onMay 22, 2025


material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
Reload insert
Tip design ART 20P

packaging

pack of 4800 ea (50 inserts of 96 tips)

manufacturer/tradename

Molecular BioProducts 2149P-05-RI

technique(s)

PCR: suitable

maximum volume

20 μL

volume

20 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service