Key Documents
Z557412
JULABO COOLING COIL FOR SHAKING WATER BATH
Select a Size
1 EA
$519.00
$519.00
Estimated to ship onApril 16, 2025
Select a Size
Change View
1 EA
$519.00
About This Item
UNSPSC Code:
41103025
NACRES:
NB.77
$519.00
Estimated to ship onApril 16, 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970416
General description
Cooling coil for retrofitting shaking water baths SW22/SW23. The cooling coil is screwed into the water bath through the drain tap. Once installed, the bath may be operated near or slightly below the ambient temperature.
Choose from one of the most recent versions:
Certificates of Analysis (COA)