Skip to Content
Merck
All Photos(2)

Key Documents

Z676446

Double-Safe PTFE coated thermometer

greener alternative

L 305 mm, temperature -20-110 °C, immersion level, 76 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$70.50

$70.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$70.50

About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

$70.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

feature

immersion level 76 mm

packaging

pack of 1 ea

manufacturer/tradename

HB Instruments B60400-0600

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

temperature measuring range

-20-110 °C

L

305 mm

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Double-Safe thermometers are PTFE coated Easy-Read thermometers with black Enviro-Safe liquid and yellow back lead free glass. Double-Safe thermometers prevent contamination of the glass and liquid in the event of breakage. Double-Safe is the ideal replacement for mercury thermometers banned (or partially banned) in many US states and most European countries.

  • PTFE coating contains liquid and glass in case of breakage
  • Non-toxic, non-hazardous, biodegradable black Enviro-Safe liquid
  • Available with statement of accuracy traceable to NIST and DKD
  • Lead-free glass and recyclable packaging
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical without mercury. Click here for more information.

Legal Information

Double-Safe is a trademark of H-B Instrument Co.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service