Key Documents
Z675776
DissecTable reversible dissecting board
polyethylene, pkg of 1 ea
Select a Size
1 EA
$885.00
$885.00
Estimated to ship onApril 17, 2025
Select a Size
Change View
1 EA
$885.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
$885.00
Estimated to ship onApril 17, 2025
material
polyethylene
packaging
pkg of 1 ea
General description
Made of heavy-duty stain resistant thick polyethylene to resist bending, or swelling. Will not dull fine surgical blades. A groove is carved all around the edge of each side of the board to contain fluids.
One side has a large cutting area including dimensional scales in inches and centimeters, along with a 60 x 80 mm grid made of 48 x 10 mm squares. Six dimensional circles are also printed from 1/8 to 5/8 in. and 4 mm to 14 mm in diameter. The other side offers 2 cutting boards half the size with the same dimensional features printed on each one. All corners have rubber feet giving more stability to the working surface.
Legal Information
DissecTable is a trademark of Simport
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. LS, What are the dimensions of this board and is it possible to autoclave this item multiple times? with regards
    1 answer
    1. The product dimensions are 575 mm x 400 mm x 12.5 mm (23 x 16 x 1/2 in). The board is made of HDPE which is not autoclavable but may be sanitized using a 70% ethanol wash.
      Helpful?