Key Documents
Z611999
Julabo® bath tubing
metal (triple insulated with connector M16x1), L 0.5 m
Select a Size
1 EA
$646.00
$646.00
Estimated to ship onMay 26, 2025
Select a Size
Change View
1 EA
$646.00
About This Item
UNSPSC Code:
41105108
NACRES:
NB.77
$646.00
Estimated to ship onMay 26, 2025
material
metal (triple insulated with connector M16x1)
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8930209
parameter
−100-350 °C temperature
L
0.5 m
General description
Flexible connection of the application to the temperature control instrument for optimal use in the respective temperature range.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)