Key Documents
Z549282
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter with return hole
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 500 mL
Select a Size
1 EA
$214.00
$214.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$214.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$214.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
female joint: ST/NS29/32
male joint: ST/NS24/40
capacity
500 mL
Related Categories
General description
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.
Application
Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)