Key Documents
Z544817
Sigma-Aldrich
Aldrich® high-efficiency variable take off head
joint: ST/NS 24/40, thermometer joint: ST/NS 10/18
Select a Size
1 EA
$1,880.00
$1,880.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,880.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$1,880.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
thermometer joint: ST/NS10/18
joint: ST/NS24/40
Related Categories
General description
Vapor from the distillation column passes through the upper holes in the take-off head and into the condenser. The condensed liquid can be drained into a side storage flask (PTFE valve open), or allowed to return to the distillation column (PTFE valve closed) through a slot in the lower part of the head. The upper and lower ST joints permit the use of take-off head with different distillation columns and condensers. Features a precision PTFE valve, and vacuum/purge hose connections with a PTFE stopcock.
A 50mm immersion length thermometer is recommended.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)