Key Documents
Z544566
Molyorbital set
Atomic Orbital
Select a Size
1 SET
$69.40
$69.40
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 SET
$69.40
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
$69.40
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
description
14-model collection set
manufacturer/tradename
Molymod
General description
Contains 1s; 2s; 2p; 3d; 2s+ 3 × 2p; and sp, sp2, and sp3 hybridized models. Pink and purple lobes represent the +/- wave phases of the p and d atomic orbitals. Models come with transparent bases for display.
The Molymod molecular orbital system uses standard shiny colored atom parts to represent the central atom cores. Average scale is 3.5 cm per Angstrom. The atomic and molecular orbital parts are represented by pastel/matte colored pieces and are color-coded according to their use.
The pink and purple pear-shaped lobes and concave π links represent the +ve and -ve signs of the wave functions Ψ of the lobes of atomic and molecular orbitals. The gray pieces represent hybridized sigma bonds. The light brown spheres are used to indicate lone electron pair orbitals. Where the electrons are involved in hydrogen bonding, a light brown pear-shaped lobe is used
Legal Information
Molyorbital is a trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)