Key Documents
Z529540
Coors porcelain pestle
for Z529516
Select a Size
1 EA
$225.00
$225.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$225.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
$225.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
porcelain
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 66332
pestle L
280 mm
General description
Oversized pestle glazed to grinding surface. Order mortar separately.
Application
The Coors porcelain pestle is suitable to use for crushing, grinding, and mixing.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)