Key Documents
Z247502
Coors porcelain pestle
for Z247464
Select a Size
1 EA
$50.50
$50.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$50.50
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
$50.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
porcelain
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 66311
pestle L
114 mm
General description
Oversized pestle glazed to grinding surface. Order mortar separately.
Application
The Coors porcelain pestle is suitable to use for crushing, grinding, and mixing.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. is the inner surface smooth or rough ?
    1 answer
    1. This item is a product of CoorsTek and includes the pestle only. This pestle is glazed or smooth up to the grinding surface.
      Helpful?