Skip to Content
Merck
All Photos(2)

Documents

Z511145

Sigma-Aldrich

Solid-phase peptide synthesis flask

capacity 1,000 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.24

sterility

non-sterile

feature

closure type Cap

packaging

pack of 1

manufacturer/tradename

Sigma-Aldrich

capacity

1,000 mL

diam. × H

115 mm × 200 mm , + - 3

Looking for similar products? Visit Product Comparison Guide

General description

Integral stirrer allows for the complete suspension of media.
Large 85 mm top and bottom openings allow for easy access when loading and cleaning.

Complete assembly includes top cap, funnel, coarse frit, stirrer assembly, 90 mm base assembly, 3 ft. of 1/4 in. PTFE tubing, 1/4 in. nut and ferrule. (Two additional ports are provided for reactant addition or gas inlet).

Features include: A removable fritted disc and a large bore 1/4-turn bottom valve for rapid filtration and washing.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service