Key Documents
Z509361
Sigma-Aldrich
Weighing funnel
size XL, anti-static PP
Synonym(s):
Measuring funnel, Scale funnel
Select a Size
10 EA
$133.00
$133.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
10 EA
$133.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.71
$133.00
Estimated to ship onMay 29, 2025
material
anti-static PP
size
XL
packaging
pkg of 10 ea
General description
  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large
Choose from one of the most recent versions:
Certificates of Analysis (COA)