Skip to Content
Merck
All Photos(1)

Key Documents

Z506575

Perfektum® one-way high pressure stopcock

male Luer lock to threaded female Luer lock

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122000
NACRES:
NB.03
Pricing and availability is not currently available.

manufacturer/tradename

Cadence Science 6021HP

General description

Chrome-plated brass body. High pressure rating up to 290psi.

Legal Information

Perfektum is a registered trademark of Cadence Science

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service