S7521
Liquid chromatography column fittings
Stopcock, 3-way Luer Lock (B)
Synonym(s):
polymeric adsorbent, 80-100 mesh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for chromatography
technique(s)
LPLC: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Nylon stopcock features 3-way control with female Luer-Lock inlet and male Luer-Lock outlet ports.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service