Key Documents
Z341797
IKA® A11 basic grinding chambers
A 11.4 grinding chamber, 1/cs
Synonym(s):
grinders, grinding chambers, grinding mills
About This Item
UNSPSC Code:
41100000
NACRES:
NB.74
feature
CE compliant
CSA approved
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 2904100
availability
not available in USA and Canada
size
250 mL , effective volume
General description
Note: Made of polycarbonate with stainless steel inlet. Not suitable for cooling with N2, only applicable with double beater A 11.6.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)