Documents
Z308900
Quickfit® conical flasks
capacity 1000 mL, joint: ST/NS 24/29
Synonym(s):
FE1L/3
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
conical flask
flat bottom
Joint
joint: ST/NS24/29
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
case of 2
manufacturer/tradename
Quickfit FE1L/3
availability
not available in USA and Canada
capacity
1000 mL
General description
With ground-glass socket for general use. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited