Skip to Content
Merck
All Photos(1)

Key Documents

Z303119

Quickfit® air condenser

L 300 mm, joint: ST/NS 24/29

Synonym(s):

FC7/23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.

Joint

joint: ST/NS24/29

manufacturer/tradename

Quickfit FC 7/23

availability

not available in USA and Canada, ROW (Available only in EU countries)
not available in USA and Canada

L

300 mm

surface area

200 cm2

Looking for similar products? Visit Product Comparison Guide

General description

These simple condensers are useful for condensation of materials with boiling points greater the 150 °C.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service