Key Documents
CLS2480300
Pyrex® Allihn condenser, with male and female joints
jacket L 300 mm, joint: ST/NS 24/40
Synonym(s):
laboratory condenser
Select a Size
1 EA
$355.00
6 EA
$1,410.00
$355.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$355.00
6 EA
$1,410.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$355.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
manufacturer/tradename
Corning 2480-300
jacket L
300 mm
Related Categories
General description
Suitable for use in vacuum distillation. With drip-tip
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)