Z265829
Preparative TLC plates
Silica gel
Synonym(s):
Prep Thin Layer Chromatography Plates
About This Item
Recommended Products
material
glass support
silica gel 60 matrix
feature
binder Organic Polymer
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
Analtech 02013
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
1,000 μm
particle size
20-40 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service