Key Documents
Z281360
Laboratory brushes
Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
Select a Size
4 EA
$73.60
$73.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EA
$73.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$73.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 4 ea
L
30 in.
brush diam. × L
1/2 in. × 3 1/4 in.
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)