Key Documents
Z274089
Sigma-Aldrich
Techware PP bench tray
pkg of 1 ea
Select a Size
1 EA
$407.00
$407.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$407.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$407.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
General description
Heavy-duty PP tray provides a solid, stable work surface. Chemically resistant and easily cleaned. Raised 1 in. lip on edges of tray provides containment of up to 2.8 L in case of spillage. Excellent for use in medium and large-size Aldrich AtmosBags as a weighted base. Built-in threaded adapter allows a 5/8 in. diam. lattice rod to be secured to tray as a vertical support for clamps. Lattice rod shown in picture not supplied with this product, purchase Z225665 separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)