Skip to Content
Merck
All Photos(2)

Key Documents

Z168777

Stainless steel forceps

Stainless steel forceps
1 of 1 reviewers received a sample product or took part in a promotion

Fine tip, straight

Synonym(s):

laboratory tweezers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$79.90

$79.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$79.90

About This Item

UNSPSC Code:
41122405
eCl@ss:
32035504
NACRES:
NB.35

$79.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 1 ea

L

4 1/4 in.

Looking for similar products? Visit Product Comparison Guide

General description

smooth spring action

Application

Stainless steel forceps are suitable for sample collection and handling.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. New Haven, CT
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Positively no complaints!

    Exactly as described, for a reasonable cost and an expedient delivery! Couldn't ask for better service.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service