Key Documents
Z265241
Supelco
TLC saturation pads
size 10 cm × 10 cm
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
packaging
pkg of 100 ea
manufacturer/tradename
Analtech 8106
technique(s)
thin layer chromatography (TLC): suitable
size
10 cm × 10 cm
General description
Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor. Pads improve reproducibility and reduce "edge effects".
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)