Key Documents
Z244120
Sigma-Aldrich
Sample cylinder plug
carbon steel
Select a Size
10 EA
$26.00
$26.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$26.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
$26.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
carbon steel
General description
The 1/4 in. NPTM square head plug fits all Sure/Pac sample cylinder sizes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)