Key Documents
Z232289
Sigma-Aldrich
Filter/Washer/Degasser
capacity 100 mL, joint: ST/NS 14/20 M, F
Select a Size
1 EA
$1,260.00
$1,260.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,260.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
$1,260.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
label
CC
Joint
joint: ST/NS14/20 M, F
capacity
100 mL
pore size
25-50 μm porosity
General description
Has septum-inlet, drip-tip, and PTFE stopcocks on side-arm, female top and male bottom joint.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.