Key Documents
Z220329
Sigma-Aldrich
Aldrich® Schlenk-type female joint cap
female joint: ST/NS 45/40
Select a Size
1 EA
$104.00
$104.00
Estimated to ship onMay 15, 2025
Select a Size
Change View
1 EA
$104.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$104.00
Estimated to ship onMay 15, 2025
label
J
K
Joint
female joint: ST/NS45/40
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.